Ethyl butyrate ≥98%, halal

Synonyms: Butanoic acid ethyl ester, Butyric acid ethyl ester

W242713-9KG-K W242713-4KG-K W242713-1KG-K W242713-20KG-K
220230-282EA 470.77 USD
220230-282 220230-281 220231-503 220231-504
Ethyl butyrate ≥98%, halal
Ethyl butyrate
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 252 °C (1013 hPa)
Melting Pt: –92 °C
Density: 0.88 g/cm³ (20 °C)
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00009394
CAS Number: 105-54-4
EINECS: 203-306-4
UN: 1180
ADR: 3,III
Merck Index: 13,03810

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR