Ethyl butyrate ≥98% Kosher

Synonyms: Butanoic acid ethyl ester, Butyric acid ethyl ester

W242705-4KG-K W242705-20KG-K W242705-1KG-K
220231-500EA 326.15 USD
220231-500 220231-499 220231-498
Ethyl butyrate ≥98% Kosher
Ethyl butyrate
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 252 °C (1013 hPa)
Melting Pt: –92 °C
Density: 0.88 g/cm³ (20 °C)
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00009394
CAS Number: 105-54-4
EINECS: 203-306-4
UN: 1180
ADR: 3,III
Merck Index: 13,03810

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR