Ethyl benzoate ≥99% Kosher

Warning

Synonyms: Benzoic acid ethyl ester

W242209-10KG-K W242209-25KG-K W242209-1KG-K
220231-486EA 450.77 USD
220231-486 220231-488 220231-487
Ethyl benzoate ≥99% Kosher
Ethyl benzoate
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 415 °C (1013 hPa)
Melting Pt: -35 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00009109
CAS Number: 93-89-0
EINECS: 202-284-3
Merck Index: 13,03802

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR