Ethyl acrylate ≥99.5% FCC

Synonyms: Acrylic acid-ethyl ester

W241806-10KG-K W241806-1KG-K W241806-25KG-K
220229-921EA 80 USD
220229-921 220231-475 220231-476
Ethyl acrylate ≥99.5% FCC
Ethyl acrylate
Formula: C₅H₈O₂
MW: 100.12 g/mol
MDL Number: MFCD00009188
CAS Number: 140-88-5
EINECS: 205-438-8
UN: 1917
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR