(±)-δ-Dodecalactone Kosher

Synonyms: 6-Heptyltetrahydro-2H-pyran-2-one, δ-Dodecalactone, 5-Dodecanolide, δ-Dodecanolactone, (±)-Tetrahydro-6-heptyl-2H-pyran-2-one, (±)-Dodecan-5-olide

W240109-5KG-K W240109-1KG-K W240109-10KG-K
220230-672EA 887.69 USD
220230-672 220230-671 220230-670
(±)-δ-Dodecalactone Kosher
(±)-δ-Dodecalactone
Formula: C₁₂H₂₂O₂
MW: 198.31 g/mol
Boiling Pt: 126…128 °C (0.5 mmHg)
Melting Pt: –7 °C
Density: 0.9585 g/cm³ (20 °C)
MDL Number: MFCD00006651
CAS Number: 713-95-1
EINECS: 211-932-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR