(±)-γ-Dodecalactone FCC

Synonyms: (±)-Dihydro-5-octylfuran-2(3H)-one, gamma-Dodecalactone, 5-Octyldihydrofuran-2(3H)-one, gamma-Dodecanolactone, gamma-Laurolactone, (±)-Dodecan-4-olide

W240001-1KG-K W240001-100G-K W240001-5KG-K
220230-667EA 227.69 USD
220230-667 220230-666 220230-668
(±)-γ-Dodecalactone FCC
(±)-γ-Dodecalactone
Formula: C₁₂H₂₂O₂
MW: 198.31 g/mol
MDL Number: MFCD00036499
CAS Number: 2305-05-7
EINECS: 218-971-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR