Dimethyl succinate ≥98%

Warning

Synonyms: DBE-4 dibasic ester, Succinic acid dimethyl ester, 1,4-Butanedioic acid dimethyl ester

W239607-10KG-K W239607-1KG-K W239607-25KG-K
220231-449EA 664.62 USD
220231-449 220231-450 220231-451
Dimethyl succinate ≥98%
Dimethyl succinate
Formula: C₆H₁₀O₄
MW: 146.14 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 18…19 °C
Density: 1.117 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008466
CAS Number: 106-65-0
EINECS: 203-419-9
Merck Index: 12,09038

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR