2,6-Dimethyl-5-heptenal FCC

Synonyms: 2,6-Dimethylhept-5-enal

W238902-1KG-K W238902-100G-K
220231-377EA 538.46 USD
220231-377 220231-376
2,6-Dimethyl-5-heptenal FCC
2,6-Dimethyl-5-heptenal
Formula: C₉H₁₆O
MW: 140.23 g/mol
MDL Number: MFCD00006981
CAS Number: 106-72-9
EINECS: 203-427-2
UN: 1989
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR