2',4'-Dimethylacetophenone ≥96% Kosher

Synonyms: 1-(2,4-Dimethylphenyl)ethan-1-one, 2,4-Dimethylacetophenone, 2,4-Dimethylacetophenon

W238708-100G-K W238708-25G-K
220231-372EA 80 USD
220231-372 220231-374
2',4'-Dimethylacetophenone ≥96% Kosher
2',4'-Dimethylacetophenone
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Boiling Pt: 70…72 °C (0.5 mmHg)
Density: 1.0121 g/cm³ (15 °C)
MDL Number: MFCD00003571
CAS Number: 89-74-7
EINECS: 201-935-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR