1,3-Dimethoxybenzene ≥98%, halal

Synonyms: Resorcinol dimethyl ether

W238503-10KG-K W238503-5KG-K W238503-1KG-K
220231-364EA 1172.31 USD
220231-364 220231-366 220231-365
1,3-Dimethoxybenzene ≥98%, halal
1,3-Dimethoxybenzene
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 218 °C (1013 hPa)
Melting Pt: –52 °C
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008384
CAS Number: 151-10-0
EINECS: 205-783-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR