Diethyl succinate ≥99%, FG

Synonyms: Succinic acid diethyl ester, 1,4-Butanedioic acid diethyl ester

W237701-10KG-K W237701-25KG-K W237701-1KG-K W237701-5KG-K
220231-339EA 583.08 USD
220231-339 220231-341 220231-340 220231-342
Diethyl succinate ≥99%, FG
Diethyl succinate
Formula: C₈H₁₄O₄
MW: 174.2 g/mol
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: –19 °C
Density: 1.04 g/cm³ (20 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00009208
CAS Number: 123-25-1
EINECS: 204-612-0
Merck Index: 12,09038

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR