Diethyl sebacate ≥98% FCC

Synonyms: Sebacic acid diethyl ester, Decanedioic acid diethyl ester, Diethyl decanedioate

W237604-1KG-K W237604-10KG-K
220231-336EA 226.15 USD
220231-336 220231-335
Diethyl sebacate ≥98% FCC
Diethyl sebacate
Formula: C₁₄H₂₆O₄
MW: 258.36 g/mol
Boiling Pt: 312 °C (1013 hPa)
Melting Pt: –3 °C
Density: 0.9646 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009219
CAS Number: 110-40-7
EINECS: 203-764-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR