Diethyl malonate ≥98%, halal

W237507-10KG W237507-1KG W237507-25KG
220231-331EA 500 USD
220231-331 220231-332 220231-333
Diethyl malonate ≥98%, halal
Diethyl malonate
Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 199 °C (1013 hPa)
Melting Pt: -50 °C
Density: 1.055 g/cm³ (20 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00009195
CAS Number: 105-53-3
EINECS: 203-305-9
Merck Index: 13,03856

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR