Decyl acetate ≥95% Kosher

Synonyms: Acetic acid decyl ester

W236705-9KG-K W236705-1KG-K W236705-4KG-K
220231-313EA 1035.38 USD
220231-313 220231-311 220231-312
Decyl acetate ≥95% Kosher
Decyl acetate
Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
MDL Number: MFCD00026538
CAS Number: 112-17-4
EINECS: 203-942-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR