trans-2-Decenal (synthetic) ≥95% FCC, FG (Halal, Kosher)

Warning

Synonyms: trans-2-Decenal, (E)-2-Decenal, (E)-Dec-2-enal

W236608-1KG-K W236608-200G-K
220230-896EA 1021.54 USD
220230-896 220230-897
trans-2-Decenal (synthetic) ≥95% FCC, FG (Halal, Kosher)
trans-2-Decenal
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 78…80 °C (3 mmHg) (lit.)
Density: 0.8572 g/cm³ (15 °C)
Flash Pt: 96 °C
MDL Number: MFCD00014679
CAS Number: 3913-81-3
EINECS: 223-472-1
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR