Capric acid ≥98%, halal

Warning

Synonyms: Decylic acid, Decanoic acid

W236411-1KG-K W236411-10KG-K W236411-5KG-K
220230-889EA 132 USD
220230-889 220230-888 220230-890
Capric acid ≥98%, halal
Capric acid
Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 29 - 32 °C
Density: 0.893 g/cm³ (20 °C)
Flash Pt: 147 °C
Storage Temperature: Ambient
MDL Number: MFCD00004441
CAS Number: 334-48-5
EINECS: 206-376-4
Merck Index: 13,01764

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR