Capric acid ≥98% Kosher

Warning

Synonyms: Decylic acid, Decanoic acid

W236403-25KG-K W236403-1KG-K W236403-5KG-K W236403-10KG-K
220230-885EA 1676.92 USD
220230-885 220230-884 220230-886 220230-883
Capric acid ≥98% Kosher
Capric acid
Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 29…32 °C
Density: 0.893 g/cm³ (20 °C)
Flash Pt: 147 °C
Storage Temperature: Ambient
MDL Number: MFCD00004441
CAS Number: 334-48-5
EINECS: 206-376-4
Merck Index: 13,01764

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR