Cyclohexylacetic acid

Synonyms: Cyclohexaneacetic acid

W234702-1KG W234702-25G W234702-100G
220231-289EA 2353.85 USD
220231-289 220231-290 220231-288
Cyclohexylacetic acid
Cyclohexylacetic acid
Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 26…27 °C
Density: 0.9985 g/cm³ (22 °C)
MDL Number: MFCD00001518
CAS Number: 5292-21-7
EINECS: 226-132-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR