4-Isopropylbenzaldehyde ≥98%

Synonyms: Cuminaldehyde

W234109-1KG-K W234109-5KG-K
220231-278EA 224.62 USD
220231-278 220231-279
4-Isopropylbenzaldehyde ≥98%
4-Isopropylbenzaldehyde
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
MDL Number: MFCD00006953
CAS Number: 122-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR