Citral diethyl acetal Kosher

W230405-100G-K W230405-1KG-K
220230-645EA 80 USD
220230-645 220230-646
Citral diethyl acetal Kosher
Citral diethyl acetal
Formula: C₁₄H₂₆O₂
MW: 226.36 g/mol
Boiling Pt: 125…130 °C (20 mmHg)
Density: 0.873 g/cm³ (20 °C)
CAS Number: 7492-66-2
EINECS: 231-323-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR