Cinnamyl alcohol ≥98%

Synonyms: Cinnamic alcohol, 3-Phenylprop-2-en-1-ol

W229407-10KG-K W229407-25KG-K W229407-5KG-K W229407-1KG-K
220231-184EA 633.85 USD
220231-184 220231-186 220231-187 220231-185
Cinnamyl alcohol ≥98%
Cinnamyl alcohol
Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 256 °C (1013 hPa)
Melting Pt: 31…34 °C
Density: 1.04 g/cm³ (25 °C)
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00002921
CAS Number: 104-54-1
EINECS: 203-212-3
Merck Index: 13,02325

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR