(R)-(-)-Carvone ≥97%, halal

Synonyms: (R)-2-Methyl-5-(prop-1-en-2-yl)cyclohex-2-enone, L-p-Mentha-1(6),8-dien-2-one, (R)-(-)-2-Methyl-5-(1-methylvinyl)cyclohex-2-en-1-one, L(-)-Carvone

W224901-1KG-K W224901-5KG-K
220230-846EA 166.15 USD
220230-846 220230-847
(R)-(-)-Carvone ≥97%, halal
(R)-(-)-Carvone
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 227 °C (1013 hPa)
Melting Pt: 25.2 °C
Density: 0.9652 g/cm³ (15 °C)
MDL Number: MFCD00001578
CAS Number: 6485-40-1
EINECS: 229-352-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR