Isobutyraldehyde ≥96%, FG

Danger

Synonyms: 2-Methylpropanal, 2-Methylpropionaldehyde

W222011-25G-K W222011-100G-K W222011-1KG-K
220230-758EA 147.54 USD
220230-758 220230-756 220230-757
Isobutyraldehyde ≥96%, FG
Isobutyraldehyde
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 64 °C (1013 hPa)
Melting Pt: –65 °C
Density: 0.794 g/cm³ (20 °C)
Flash Pt: –24 °C
Storage Temperature: Ambient
MDL Number: MFCD00006980
CAS Number: 78-84-2
EINECS: 201-149-6
UN: 2045
ADR: 3,II
Merck Index: 13,05171

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR