Isobutyraldehyde ≥98% Kosher

Danger

Synonyms: 2-Methylpropanal, 2-Methylpropionaldehyde

W222003-1KG-K W222003-4KG-K W222003-8KG-K W222003-20KG-K
220230-751EA 130 USD
220230-751 220230-753 220230-754 220230-752
Isobutyraldehyde ≥98% Kosher
Isobutyraldehyde
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 64 °C (1013 hPa)
Melting Pt: –65 °C
Density: 0.794 g/cm³ (20 °C)
Flash Pt: –24 °C
Storage Temperature: Ambient
MDL Number: MFCD00006980
CAS Number: 78-84-2
EINECS: 201-149-6
UN: 2045
ADR: 3,II
Merck Index: 13,05171

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR