Butyl isovalerate ≥98%, halal

Synonyms: Isovaleric acid butyl ester, Butyl 3-methylbutanoate

W221805-1KG-K W221805-20KG-K W221805-4KG-K
220231-042EA 114.46 USD
220231-042 220231-043 220230-744
Butyl isovalerate ≥98%, halal
Butyl isovalerate
Formula: C₉H₁₈O₂
MW: 158.24 g/mol
MDL Number: MFCD00048775
CAS Number: 109-19-3
EINECS: 203-654-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR