Dibutyl sulfide ≥95%

Synonyms: Dibutylsulphide, Butyl sulphide, n-Butyl sulphide

W221503-8KG-K W221503-4KG-K W221503-1KG-K
220231-032EA 1176.92 USD
220231-032 220231-031 220231-030
Dibutyl sulfide ≥95%
Dibutyl sulphide
Formula: C₈H₁₈S
MW: 146.3 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: –80 °C
Density: 0.83 g/cm³ (20 °C)
Flash Pt: 62 °C
MDL Number: MFCD00009468
CAS Number: 544-40-1
EINECS: 208-870-5
UN: 3334
ADR: 9,
Merck Index: 12,01626

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR