Isobutyl isobutyrate ≥97%

W218901-4KG-K W218901-1KG-K
220230-951EA 350.77 USD
220230-951 220230-950
Isobutyl isobutyrate ≥97%
Isobutyl isobutyrate
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
MDL Number: MFCD00008916
CAS Number: 97-85-8
EINECS: 202-612-5
UN: 2528
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR