Butyl isobutyrate ≥97%

Synonyms: Isobutyric acid butyl ester

W218804-5KG-K W218804-9KG-K W218804-1KG-K
220230-947EA 729.23 USD
220230-947 220230-948 220230-946
Butyl isobutyrate ≥97%
Butyl isobutyrate
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
MDL Number: MFCD00048773
CAS Number: 97-87-0
EINECS: 202-614-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR