Butyl butyrate ≥98% FCC

Danger

Synonyms: Butyric acid butyl ester

W218618-9KG-K W218618-1KG-K
220230-932EA 333.85 USD
220230-932 220230-930
Butyl butyrate ≥98% FCC
Butyl butyrate
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 166 °C (1013 hPa)
Melting Pt: –91.5 °C
Density: 0.872 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009450
CAS Number: 109-21-7
EINECS: 203-656-8
UN: 3272
ADR: 3,III
Merck Index: 13,01555

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR