2,6-Di-tert-butyl-p-cresol ≥99% FCC

Synonyms: Butylated hydroxytoluene, Butylhydroxytoluene, 2,6-Di-tert-butyl-4-methylphenol, BHT

W218405-10KG-K W218405-1KG-K
220230-922EA 607.69 USD
220230-922 220230-923
2,6-Di-tert-butyl-p-cresol ≥99% FCC
2,6-Di-tert-butyl-p-cresol
Formula: C₁₅H₂₄O
MW: 220.35 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 69 - 70 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00011644
CAS Number: 128-37-0
EINECS: 204-881-4
Merck Index: 13,01547

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR