tert-Butyl-4-methoxyphenol (mixture of isomers) ≥98.5%, halal

Synonyms: tert-Butyl-4-methoxyphenol, BHA, tert-Butyl-4-hydroxyanisole, Butylhydroxyanisole, 2(3)-tert-Butyl-4-methoxyphenol, Butylated hydroxyanisole

W218308-5KG-K W218308-100G-K W218308-1KG-K
220230-920EA 580 USD
220230-920 220230-918 220230-919
tert-Butyl-4-methoxyphenol (mixture of isomers) ≥98.5%, halal
tert-Butyl-4-methoxyphenol (mixture of isomers)
Formula: C₁₁H₁₆O₂
MW: 180.25 g/mol
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 48…55 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD01779059
CAS Number: 25013-16-5
EINECS: 246-563-8
Merck Index: 12,01582

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR