Benzyl cinnamate

Synonyms: Cinnamic acid benzyl ester, trans-Cinnamic acid benzyl ester, 3-Phenyl-2-propenoic acid benzyl ester

W214205-10KG-K W214205-1KG-K W214205-5KG-K
220230-635EA 1530.77 USD
220230-635 220230-636 220230-637
Benzyl cinnamate
Benzyl cinnamate
Formula: C₁₆H₁₄O₂
MW: 238.29 g/mol
MDL Number: MFCD00004789
CAS Number: 103-41-3
EINECS: 203-109-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR