Benzyl isobutyrate, halal

W214108-10KG-K W214108-1KG-K W214108-5KG-K
220229-698EA 612.31 USD
220229-698 220229-699 220229-700
Benzyl isobutyrate, halal
Benzyl isobutyrate
Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
MDL Number: MFCD00048315
CAS Number: 103-28-6
EINECS: 203-095-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR