Benzyl butyrate ≥98% Kosher

Synonyms: Butyric acid benzyl ester, Butanoic acid benzyl ester

W214019-100G-K W214019-1KG-K
220230-633EA 189.23 USD
220230-633 220229-694
Benzyl butyrate ≥98% Kosher
Benzyl butyrate
Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 240 °C (1013 hPa)
Density: 1.01 g/cm³ (20 °C)
MDL Number: MFCD00027133
CAS Number: 103-37-7
EINECS: 203-105-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR