Benzyl butyrate ≥98%, halal

Synonyms: Butyric acid benzyl ester, Butanoic acid benzyl ester

W214000-10KG-K W214000-1KG-K W214000-5KG-K
220230-629EA 749.23 USD
220230-629 220230-630 220230-631
Benzyl butyrate ≥98%, halal
Benzyl butyrate
Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 240 °C (1013 hPa)
Density: 1.01 g/cm³ (20 °C)
MDL Number: MFCD00027133
CAS Number: 103-37-7
EINECS: 203-105-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR