Isopentyl hexanoate mixture of isomers ≥98% Kosher

Synonyms: Isopentyl hexanoate, Isoamyl caproate, Isoamyl hexanoate, Hexanoic acid Isopentyl ester, Hexanoic acid isoamyl ester, Isopentyl caproate, Caproic acid isoamyl ester, Caproic acid isopentyl ester, 3-Methylbutyl hexanoate, Hexanoic acid isopentyl ester

W207519-4KG-K W207519-100G-K W207519-1KG-K
220230-465EA 80 USD
220230-465 220230-463 220230-464
Isopentyl hexanoate mixture of isomers ≥98% Kosher
Isopentyl hexanoate
Formula: C₁₁H₂₂O₂
MW: 186.29 g/mol
Boiling Pt: 226 °C (1013 hPa)
Density: 0.861 g/cm³ (20 °C)
MDL Number: MFCD00027280
CAS Number: 2198-61-0
EINECS: 218-600-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR