Diallyl sulfide ≥97%, FG

Synonyms: Allyl sulphide

W204218-1KG-K W204218-100G-K W204218-4KG-K
220230-392EA 660 USD
220230-392 220230-391 220230-393
Diallyl sulfide ≥97%, FG
Diallyl sulphide
Formula: C₆H₁₀S
MW: 114.21 g/mol
Boiling Pt: 139…140 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008658
CAS Number: 592-88-1
EINECS: 209-775-1
UN: 1993
ADR: 3,III
Merck Index: 13,00294

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR