Diallyl disulfide ≥80% Kosher

Warning

Synonyms: Diallyldisulphide, Allyl disulphide

W202800-5KG-K W202800-100G-K W202800-1KG-K
220230-347EA 1892.31 USD
220230-347 220230-345 220230-346
Diallyl disulfide ≥80% Kosher
Diallyl disulphide
Formula: C₆H₁₀S₂
MW: 146.28 g/mol
Boiling Pt: 198 °C (1013 hPa)
Density: 1.0237 g/cm³ (15 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00008656
CAS Number: 2179-57-9
EINECS: 218-548-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR