Sodium alginate

Synonyms: Alginic acid sodium salt, Algin

W201502-1KG W201502-5KG
220230-322EA 230.77 USD
220230-322 220230-323
Sodium alginate
Sodium alginate
Storage Temperature: Ambient
MDL Number: MFCD00081310
CAS Number: 9005-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR