Acetophenone ≥98% Kosher

Warning

Synonyms: Methyl phenyl ketone, Phenyl methyl ketone

W200905-10KG-K W200905-1KG-K W200905-25KG-K
220229-823EA 510.77 USD
220229-823 220229-824 220229-825
Acetophenone ≥98% Kosher
Acetophenone
Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 201.7 °C (1013 hPa)
Melting Pt: 20 °C
Density: 1.027 g/cm³ (20 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00008724
CAS Number: 98-86-2
EINECS: 202-708-7
UN: 3334
ADR: 9,
Merck Index: 13,00074

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR