Triacetin (glycerol triacetate) ≥98.5%

Synonyms: Glyceryl triacetate, Propane-1,2,3-triyl triacetate, Glycerol triacetate

W200700-1KG-K W200700-25KG-K W200700-10KG-K
220227-487EA 117.69 USD
220227-487 220227-488 220227-486
Triacetin (glycerol triacetate) ≥98.5%
Triacetin (glycerol triacetate)
Formula: C₉H₁₄O₆
MW: 218.21 g/mol
Boiling Pt: 258 °C (1013 hPa)
Melting Pt: 4 °C
Density: 1.1596 g/cm³ (20 °C)
Flash Pt: 138 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008716
CAS Number: 102-76-1
EINECS: 203-051-9
Merck Index: 13,09664

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR