Valerophenone ≥99%

Synonyms: 1-Phenyl-1-pentanone, 1-Phenylpentan-1-one

V659-10G V659-25G V659-100G
220230-066EA 35.54 USD
220230-066 220230-067 220230-065
Valerophenone ≥99%
Valerophenone
Formula: C₆H₅C(O)(CH₂)₃CH₃
MW: 162.23 g/mol
Boiling Pt: 249 °C (1013 hPa)
Melting Pt: -9 °C
Density: 0.98777 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009480
CAS Number: 1009-14-9
EINECS: 213-767-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2584f16b192b481fbaf1d8ab1e77be6109fb67a6&vid=3bf6c22512cdad990d29bc1ffe3db3b535a55970&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR