(±)-γ-Valerolactone

Synonyms: (±)-4-Hydroxypentanoic acid lactone, (±)-Pentan-4-olide, γ-Pentanolactone, 5-Methyldihydrofuran-2(3H)-one, gamma-Valerolactone

V403-100G V403-500G
220229-647EA 73.77 USD
220229-647 220229-648
(±)-γ-Valerolactone
(±)-γ-Valerolactone
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 207 °C (1013 hPa)
Melting Pt: –31 °C
Density: 1.05 g/cm³ (20 °C)
MDL Number: MFCD00005400
CAS Number: 108-29-2
EINECS: 203-569-5

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR