4-Vinylpyridine

V3204-5ML V3204-100ML V3204-500ML
220229-463EA 34.26 USD
220229-463 220229-461 220229-462
4-Vinylpyridine
4-Vinylpyridine
Formula: C₇H₇N
MW: 105.14 g/mol
Boiling Pt: 121 °C (1013 hPa)
Melting Pt: –39 °C
Density: 0.98 g/cm³ (20 °C)
Flash Pt: 48 °C
Storage Temperature: Freezer
MDL Number: MFCD00006447
CAS Number: 100-43-6
EINECS: 202-852-0
UN: 3073
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR