3,4-Dimethoxybenzylamine ≥97%

Synonyms: Veratrylamine

V1309-5G V1309-25G
220229-798EA 70 USD
220229-798 220229-797
3,4-Dimethoxybenzylamine ≥97%
3,4-Dimethoxybenzylamine
Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
Boiling Pt: 154…158 °C (12 mmHg)
Density: 1.109 g/cm³ (20 °C)
MDL Number: MFCD00008116
CAS Number: 5763-61-1
EINECS: 227-287-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR