Uric acid

U2625-25G U2625-1KG U2625-100G
220226-302EA 68.77 USD
220226-302 220226-301 220226-300
Uric acid
Uric acid
Formula: C₅H₄N₄O₃
Melting Pt: 436 °C
Density: 1.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005712
CAS Number: 69-93-2
EINECS: 200-720-7
Merck Index: 12,10014

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR