10-Undecen-1-ol ≥98%

Synonyms: 1-Undecen-11-ol, 10-Undecen-1-ol

U2008-25G U2008-100G
220227-591EA 71.83 USD
220227-591 220229-252
10-Undecen-1-ol ≥98%
10-Undecen-1-ol
Formula: C₁₁H₂₂O
MW: 170.3 g/mol
Boiling Pt: 75…80 °C (0.05 mmHg)
Melting Pt: –1 °C
Density: 0.86 g/cm³ (0 °C)
MDL Number: MFCD00004750
CAS Number: 112-43-6
EINECS: 203-971-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR