2-Undecanone ≥99%

Synonyms: Methyl nonyl ketone, Nonyl methyl ketone

U1303-250ML U1303-5ML
220229-234EA 86.91 USD
220229-234 220229-235
2-Undecanone ≥99%
2-Undecanone
Formula: C₁₁H₂₂O
MW: 170.3 g/mol
Boiling Pt: 231 °C (1013 hPa)
Melting Pt: ∼12 °C
Density: 0.83 g/cm³ (20 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00009583
CAS Number: 112-12-9
EINECS: 203-937-5
UN: 3082
ADR: 9,III
Merck Index: 12,06178

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR