Methyl undecanoate ≥99% (by GC)

Synonyms: Undecanoic acid methyl ester, Undecylic acid methyl ester

U0250-10G U0250-25G U0250-5G
220229-225EA 163.69 USD
220229-225 220229-226 220228-650
Methyl undecanoate ≥99% (by GC)
Methyl undecanoate
Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
MDL Number: MFCD00008957
CAS Number: 1731-86-8
EINECS: 217-053-2

Order Now

Specification Test Results

Assay ~99 % (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR