Thiourea ≥99.0% ACS

Warning

T8656-500G T8656-50G T8656-100G
220228-140EA 113.08 USD
220228-140 220228-141 220228-139
Thiourea ≥99.0% ACS
Thiourea
Formula: NH₂CSNH₂
Melting Pt: 174…177 °C
Density: 1.435 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008067
CAS Number: 62-56-6
EINECS: 200-543-5
UN: 3077
ADR: 9,III
Merck Index: 13,09443
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fff6222c54fe16ea4d36edcbf320745cb1e18d6&vid=e9ae5dc49e92d68f893244a5cd4e0245f289ed09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR